5-(2-Methoxy-4-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
Supplier: AOBChem USA
|
Warning
|
59541-1G
59541-25G
59541-500MG
59541-5G
76934-968EA
441.34
USD
76934-968
76934-970
76934-972
76934-974
5-(2-Methoxy-4-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Methoxy-4-(trifluoromethyl)phenyl)thiazol-2-amine
Precaución: for Research Use Only
Formula:
C₁₁H₉F₃N₂OS MW: 274.262 g/mol Storage Temperature: Ambient |
CAS Number:
2755724-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...