5-(2-Chloro-4-methoxyphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

59539-1G 59539-500MG 59539-5G
76934-956EA 494.38 USD
76934-956 76934-958 76934-960
5-(2-Chloro-4-methoxyphenyl)thiazol-2-amine ≥95%
5-(2-Chloro-4-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂OS
MW: 240.709 g/mol
Storage Temperature: Ambient
CAS Number: 2755719-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR