5-(3-Ethoxy-5-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59537-1G 59537-500MG 59537-5G
76934-942EA 589.87 USD
76934-942 76934-944 76934-946
5-(3-Ethoxy-5-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Ethoxy-5-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁FN₂OS
MW: 238.281 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-19-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR