5-(5-Chloro-2-ethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59535-1G 59535-500MG 59535-5G
76934-928EA 671.55 USD
76934-928 76934-930 76934-932
5-(5-Chloro-2-ethoxyphenyl)thiazol-2-amine ≥97%
5-(5-Chloro-2-ethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂OS
MW: 254.736 g/mol
Storage Temperature: Refrigerator
CAS Number: 2755718-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR