5-(3-Chloro-2-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59418-1G 59418-500MG 59418-5G
76934-876EA 553.79 USD
76934-876 76934-878 76934-880
5-(3-Chloro-2-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(3-Chloro-2-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClN₂OS
Storage Temperature: Refrigerator
CAS Number: 2755724-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR