5-(4-Butoxy-3-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59409-1G 59409-25G 59409-500MG 59409-5G
76934-844EA 296 USD
76934-844 76934-846 76934-848 76934-850
5-(4-Butoxy-3-fluorophenyl)thiazol-2-amine ≥97%
5-(4-Butoxy-3-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₅FN₂OS
MW: 266.334 g/mol
Storage Temperature: Ambient
CAS Number: 2807466-06-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR