5-(4-Isopropoxy-2-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59407-1G 59407-500MG 59407-5G
76934-832EA 702.32 USD
76934-832 76934-834 76934-836
5-(4-Isopropoxy-2-methoxyphenyl)thiazol-2-amine ≥97%
5-(4-Isopropoxy-2-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂O₂S
MW: 264.343 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR