5-(3-Fluoro-4-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59397-1G 59397-500MG 59397-5G
76934-780EA 524.09 USD
76934-780 76934-782 76934-784
5-(3-Fluoro-4-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-4-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.308 g/mol
Storage Temperature: Ambient
CAS Number: 2755717-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR