4-Chloro-2,5-dimethylphenol ≥97%

Supplier: AOBChem USA

Warning

59002-1G 59002-25G 59002-500MG 59002-5G
76934-746EA 241.89 USD
76934-746 76934-748 76934-750 76934-752
4-Chloro-2,5-dimethylphenol ≥97%
4-Chloro-2,5-dimethylphenol

Precaución: for Research Use Only

Formula: C₈H₉ClO
Storage Temperature: Ambient
CAS Number: 1124-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR