5-(2-Fluoro-5-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58894-1G 58894-500MG 58894-5G
76934-734EA 524.09 USD
76934-734 76934-736 76934-738
5-(2-Fluoro-5-methoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-5-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂OS
MW: 224.255 g/mol
Storage Temperature: Ambient
CAS Number: 2755722-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR