5-(4-Fluoro-3-isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58890-1G 58890-500MG 58890-5G
76934-710EA 664.13 USD
76934-710 76934-712 76934-714
5-(4-Fluoro-3-isopropoxyphenyl)thiazol-2-amine ≥97%
5-(4-Fluoro-3-isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₃FN₂OS
MW: 252.308 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-08-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR