5-(3-Chloro-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58883-1G 58883-500MG 58883-5G
76934-674EA 494.38 USD
76934-674 76934-676 76934-678
5-(3-Chloro-2-methylphenyl)thiazol-2-amine ≥97%
5-(3-Chloro-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂S
MW: 224.71 g/mol
Storage Temperature: Ambient
CAS Number: 2755724-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR