5-(2-Chloro-4-ethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58881-1G 58881-500MG 58881-5G
76934-660EA 500.75 USD
76934-660 76934-662 76934-664
5-(2-Chloro-4-ethoxyphenyl)thiazol-2-amine ≥97%
5-(2-Chloro-4-ethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂OS
MW: 254.736 g/mol
Storage Temperature: Ambient
CAS Number: 2764729-42-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR