5-(3-Bromo-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58875-1G 58875-500MG 58875-5G
76934-634EA 678.98 USD
76934-634 76934-636 76934-638
5-(3-Bromo-2-methylphenyl)thiazol-2-amine ≥97%
5-(3-Bromo-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂S
MW: 269.161 g/mol
Storage Temperature: Ambient
CAS Number: 2755721-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR