5-(4-(Benzyloxy)-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58682-1G 58682-25G 58682-500MG 58682-5G
76934-590EA 434.97 USD
76934-590 76934-592 76934-594 76934-596
5-(4-(Benzyloxy)-2-methylphenyl)thiazol-2-amine ≥97%
5-(4-(Benzyloxy)-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂OS
MW: 296.387 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-91-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR