5-(5-(Benzyloxy)-2-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58680-1G 58680-500MG 58680-5G
76934-578EA 494.38 USD
76934-578 76934-580 76934-582
5-(5-(Benzyloxy)-2-methylphenyl)thiazol-2-amine ≥97%
5-(5-(Benzyloxy)-2-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂OS
MW: 296.387 g/mol
Storage Temperature: Ambient
CAS Number: 2764731-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR