5-(3-(Benzyloxy)-4-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58678-1G 58678-500MG 58678-5G
76934-566EA 524.09 USD
76934-566 76934-568 76934-570
5-(3-(Benzyloxy)-4-fluorophenyl)thiazol-2-amine ≥97%
5-(3-(Benzyloxy)-4-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₃FN₂OS
MW: 300.351 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-37-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR