5-(3-Fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58289-1G 58289-500MG 58289-5G
76934-518EA 553.79 USD
76934-518 76934-520 76934-522
5-(3-Fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆F₄N₂S
MW: 262.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27441620
CAS Number: 1712432-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR