5-(4-Bromo-2-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58285-1G 58285-500MG 58285-5G
76934-506EA 525.15 USD
76934-506 76934-508 76934-510
5-(4-Bromo-2-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2-(trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂OS
MW: 339.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564548
CAS Number: 2734775-38-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR