5-(2-(Benzyloxy)-5-methylphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

58283-1G 58283-500MG 58283-5G
76934-494EA 524.09 USD
76934-494 76934-496 76934-498
5-(2-(Benzyloxy)-5-methylphenyl)thiazol-2-amine ≥95%
5-(2-(Benzyloxy)-5-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂OS
MW: 296.387 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564546
CAS Number: 2734778-17-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR