5-(2-Fluoro-6-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58281-1G 58281-500MG 58281-5G
76934-482EA 525.15 USD
76934-482 76934-484 76934-486
5-(2-Fluoro-6-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-6-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆F₄N₂S
MW: 262.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564545
CAS Number: 2734776-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR