5-(2-Bromo-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57680-1G 57680-500MG 57680-5G
76934-454EA 678.98 USD
76934-454 76934-456 76934-458
5-(2-Bromo-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Bromo-3-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂S
MW: 323.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564481
CAS Number: 2734773-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR