5-(2-Fluoro-3-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57678-1G 57678-500MG 57678-5G
76934-442EA 500.75 USD
76934-442 76934-444 76934-446
5-(2-Fluoro-3-methoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-3-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂OS
MW: 224.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD24308772
CAS Number: 1550673-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR