5-Ethyl-2-methoxybenzaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

57607-1G 57607-25G 57607-500MG 57607-5G
76934-428EA 263.11 USD
76934-428 76934-430 76934-432 76934-434
5-Ethyl-2-methoxybenzaldehyde ≥97% research grade
5-Ethyl-2-methoxybenzaldehyde
Formula: C₁₀H₁₂O₂
Storage Temperature: Ambient
MDL Number: MFCD06247461
CAS Number: 85944-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR