5-(2-Chloro-4-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57437-1G 57437-500MG 57437-5G
76934-392EA 500.75 USD
76934-392 76934-394 76934-396
5-(2-Chloro-4-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Chloro-4-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆ClF₃N₂S
MW: 278.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530594
CAS Number: 2734777-48-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR