5-(2-(Benzyloxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57435-1G 57435-25G 57435-500MG 57435-5G
76934-378EA 441.34 USD
76934-378 76934-380 76934-382 76934-384
5-(2-(Benzyloxy)phenyl)thiazol-2-amine ≥97%
5-(2-(Benzyloxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₄N₂OS
Storage Temperature: Ambient
MDL Number: MFCD34564443
CAS Number: 2288345-11-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR