5-(2,6-Difluoro-4-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57403-1G 57403-500MG 57403-5G
76934-366EA 631.24 USD
76934-366 76934-368 76934-370
5-(2,6-Difluoro-4-methoxyphenyl)thiazol-2-amine ≥97%
5-(2,6-Difluoro-4-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₂N₂OS
MW: 242.245 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12779886
CAS Number: 1183701-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR