5-(2-Bromo-4-(trifluoromethoxy)phenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

57174-1G 57174-500MG 57174-5G
76934-346EA 589.87 USD
76934-346 76934-348 76934-350
5-(2-Bromo-4-(trifluoromethoxy)phenyl)thiazol-2-amine ≥95%
5-(2-Bromo-4-(trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂OS
Storage Temperature: Ambient
MDL Number: MFCD34530581
CAS Number: 2734774-13-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR