5-(3-(Trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57171-1G 57171-25G 57171-500MG 57171-5G
76934-330EA 434.97 USD
76934-330 76934-332 76934-334 76934-336
5-(3-(Trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(3-(Trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂OS
MW: 260.236 g/mol
Storage Temperature: Ambient
CAS Number: 499240-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR