5-Chloro-1,3-difluoro-2-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

56943-1G 56943-25G 56943-500MG 56943-5G
76934-302EA 215.37 USD
76934-302 76934-304 76934-306 76934-308
5-Chloro-1,3-difluoro-2-iodobenzene ≥97%
5-Chloro-1,3-difluoro-2-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₂ClF₂I
Storage Temperature: Ambient
MDL Number: MFCD12922625
CAS Number: 1208076-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR