5-(3-Bromo-5-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56622-1G 56622-25G 56622-500MG 56622-5G
76934-286EA 393.6 USD
76934-286 76934-288 76934-290 76934-292
5-(3-Bromo-5-methylphenyl)thiazol-2-amine ≥97%
5-(3-Bromo-5-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂S
MW: 269.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564397
CAS Number: 2734779-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR