5-(4-Bromo-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56618-1G 56618-500MG 56618-5G
76934-262EA 560.16 USD
76934-262 76934-264 76934-266
5-(4-Bromo-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(4-Bromo-3-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂S
MW: 323.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564396
CAS Number: 2734778-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR