5-(2-Bromo-5-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56614-1G 56614-25G 56614-500MG 56614-5G
76934-234EA 428.61 USD
76934-234 76934-236 76934-238 76934-240
5-(2-Bromo-5-methylphenyl)thiazol-2-amine ≥97%
5-(2-Bromo-5-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂S
MW: 269.161 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564394
CAS Number: 2734777-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR