5-(2-Fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56612-1G 56612-500MG 56612-5G
76934-222EA 493.32 USD
76934-222 76934-224 76934-226
5-(2-Fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-5-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆F₄N₂S
MW: 262.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564392
CAS Number: 2734773-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR