5-(5-Fluoro-2-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56609-1G 56609-25G 56609-500MG 56609-5G
76934-208EA 381.93 USD
76934-208 76934-210 76934-212 76934-214
5-(5-Fluoro-2-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(5-Fluoro-2-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆F₄N₂S
MW: 262.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564391
CAS Number: 2734778-99-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR