5-(4-Bromo-2,6-difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56241-1G 56241-25G 56241-500MG 56241-5G
76934-180EA 375.56 USD
76934-180 76934-182 76934-184 76934-186
5-(4-Bromo-2,6-difluorophenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2,6-difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅BrF₂N₂S
Storage Temperature: Ambient
CAS Number: 2002352-15-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR