5-(2-Fluoro-4,5-dimethylphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

56219-1G 56219-500MG 56219-5G
76934-168EA 732.03 USD
76934-168 76934-170 76934-172
5-(2-Fluoro-4,5-dimethylphenyl)thiazol-2-amine ≥95%
5-(2-Fluoro-4,5-dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁FN₂S
MW: 222.282 g/mol
Storage Temperature: Ambient
CAS Number: 1866932-76-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR