5-(2,6-Difluoro-3-methylphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

56152-1G 56152-500MG 56152-5G
76934-154EA 523.03 USD
76934-154 76934-156 76934-158
5-(2,6-Difluoro-3-methylphenyl)thiazol-2-amine ≥95%
5-(2,6-Difluoro-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈F₂N₂S
MW: 226.246 g/mol
Storage Temperature: Ambient
CAS Number: 1556200-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR