5-(3-Fluoro-5-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56136-1G 56136-25G 56136-500MG 56136-5G
76934-140EA 441.34 USD
76934-140 76934-142 76934-144 76934-146
5-(3-Fluoro-5-methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-5-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂OS
MW: 224.255 g/mol
Storage Temperature: Ambient
CAS Number: 2007405-27-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR