5-(3-Fluoro-4-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56109-1G 56109-25G 56109-500MG 56109-5G
76934-124EA 296 USD
76934-124 76934-126 76934-128 76934-130
5-(3-Fluoro-4-methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Fluoro-4-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂OS
MW: 224.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12824181
CAS Number: 1178262-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR