5-(2,5-Dibromophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56060-1G 56060-500MG 56060-5G
76934-110EA 495.45 USD
76934-110 76934-112 76934-114
5-(2,5-Dibromophenyl)thiazol-2-amine ≥97%
5-(2,5-Dibromophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆Br₂N₂S
Storage Temperature: Ambient
CAS Number: 2290423-09-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR