5-(2-Bromo-5-chlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56046-1G 56046-500MG 56046-5G
76934-098EA 494.38 USD
76934-098 76934-100 76934-102
5-(2-Bromo-5-chlorophenyl)thiazol-2-amine ≥97%
5-(2-Bromo-5-chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrClN₂S
MW: 289.579 g/mol
Storage Temperature: Ambient
CAS Number: 1599301-73-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR