5-(2-Isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56023-1G 56023-500MG 56023-5G
76934-086EA 527.27 USD
76934-086 76934-088 76934-090
5-(2-Isopropoxyphenyl)thiazol-2-amine ≥97%
5-(2-Isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂OS
MW: 234.317 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23895300
CAS Number: 1501096-89-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR