5-(2,5-Difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56002-1G 56002-25G 56002-500MG 56002-5G
76934-072EA 406.33 USD
76934-072 76934-074 76934-076 76934-078
5-(2,5-Difluorophenyl)thiazol-2-amine ≥97%
5-(2,5-Difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆F₂N₂S
MW: 212.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12824114
CAS Number: 1179312-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR