5-(3-Bromo-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55970-1G 55970-25G 55970-500MG 55970-5G
76934-048EA 372.38 USD
76934-048 76934-050 76934-052 76934-054
5-(3-Bromo-2-fluorophenyl)thiazol-2-amine ≥97%
5-(3-Bromo-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂S
MW: 273.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564330
CAS Number: 1692422-01-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR