5-(4-Bromo-2-chlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55957-1G 55957-25G 55957-500MG 55957-5G
76934-032EA 381.93 USD
76934-032 76934-034 76934-036 76934-038
5-(4-Bromo-2-chlorophenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2-chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrClN₂S
Storage Temperature: Ambient
MDL Number: MFCD34530465
CAS Number: 1592627-46-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR