5-(2,3-Difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55954-1G 55954-25G 55954-500MG 55954-5G
76934-016EA 405.27 USD
76934-016 76934-018 76934-020 76934-022
5-(2,3-Difluorophenyl)thiazol-2-amine ≥97%
5-(2,3-Difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆F₂N₂S
MW: 212.219 g/mol
Storage Temperature: Ambient
MDL Number: MFCD19626862
CAS Number: 1340417-36-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR