5-(2,3-Dihydrobenzofuran-5-yl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

55938-1G 55938-500MG 55938-5G
76934-002EA 560.16 USD
76934-002 76934-004 76934-006
5-(2,3-Dihydrobenzofuran-5-yl)thiazol-2-amine ≥95%
5-(2,3-Dihydrobenzofuran-5-yl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂OS
Storage Temperature: Ambient
CAS Number: 1179188-58-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR