5-(2-Chloro-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55909-1G 55909-500MG 55909-5G
76933-990EA 500.75 USD
76933-990 76933-992 76933-994
5-(2-Chloro-4-methylphenyl)thiazol-2-amine ≥97%
5-(2-Chloro-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂S
MW: 224.71 g/mol
Storage Temperature: Ambient
CAS Number: 2025040-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR