5-(4-Fluoronaphthalen-1-yl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55858-1G 55858-500MG 55858-5G
76933-954EA 500.75 USD
76933-954 76933-956 76933-958
5-(4-Fluoronaphthalen-1-yl)thiazol-2-amine ≥97%
5-(4-Fluoronaphthalen-1-yl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₉FN₂S
MW: 244.287 g/mol
Storage Temperature: Ambient
CAS Number: 1486890-74-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR